Learn More
Medchemexpress LLC HY-14802D 1mg Medchemexpress, (-)-Talarozole CAS:201410-67-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY14802D1MG
Encompass_Preferred
Medchemexpress, HY-14802D 1mg (-)-Talarozole CAS:201410-67-5 (-)-Talarozole is a potent inhibitor of retinoic acid metabolism extracted from patent WO 1997049704 A1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2707
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.