Learn More
Medchemexpress LLC HY-14802C 5mg Medchemexpress, (+)-Talarozole CAS:201410-66-4 Purity:>98%
SDP
Catalog No. 502081641
Encompass_Preferred
Medchemexpress, HY-14802C 5mg (+)-Talarozole CAS:201410-66-4 (+)-Talarozole is a potent inhibitor of retinoic acid metabolism extracted from patent WO 1997049704 A1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1641 Supplier Medchemexpress LLC Supplier No. HY14802C5MG
Medchemexpress, HY-14802C 5mg (+)-Talarozole CAS:201410-66-4 (+)-Talarozole is a potent inhibitor of retinoic acid metabolism extracted from patent WO 1997049704 A1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.