Learn More
Cayman Chemical Talatisamine, Purity: ≥98%, CAS: 20501-56-8, 50 mg
SDP
Catalog No. 502939173
Encompass_Preferred
A diterpene alkaloid with diverse biological activities; blocks IK current in rat hippocampal neurons (IC50 = 146 μM); reduces Aβ40-induced increases in IK current and cytotoxicity in primary rat cortical neurons; inhibits barium chloride-induced contractions in isolated rat intenstines (EC50 = 200 μM); exhibits feeding deterrent activity against red flour beetle (T. castaneum) adults (EC50 = 342.8 ppm)
Catalog No. 50-293-9173 Supplier Cayman Chemical Supplier No. 2772150 MG
A diterpene alkaloid with diverse biological activities; blocks IK current in rat hippocampal neurons (IC50 = 146 μM); reduces Aβ40-induced increases in IK current and cytotoxicity in primary rat cortical neurons; inhibits barium chloride-induced contractions in isolated rat intenstines (EC50 = 200 μM); exhibits feeding deterrent activity against red flour beetle (T. castaneum) adults (EC50 = 342.8 ppm)