Learn More
Medchemexpress LLC HY-16106 50mg Medchemexpress, Talazoparib CAS:1207456-01-6 Purity:>98%
SDP
Catalog No. 502029189
Encompass_Preferred
Medchemexpress, HY-16106 50mg Talazoparib CAS:1207456-01-6 Talazoparib (BMN-673) is a highly potent PARP1/2 inhibitor with Kis of 1.2 nM and 0.87 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9189 Supplier Medchemexpress LLC Supplier No. HY1610650MG
Medchemexpress, HY-16106 50mg Talazoparib CAS:1207456-01-6 Talazoparib (BMN-673) is a highly potent PARP1/2 inhibitor with Kis of 1.2 nM and 0.87 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.