Learn More
Troemner™ Talboys™ 2-Prong Dual Adjust Nickel-Plated Zinc Clamp
Catalog No. 02216209 Shop All Troemner Products
Change view
Click to view available options
Material:
Nickel-plated Zinc
Catalog No. Material
02-216-209 Nickel-plated Zinc
1 options
Catalog No. 02-216-209 Supplier Troemner™ Supplier No. 30392207
Add to Cart
Add to Cart
Designed to securely hold laboratory glassware and apparatus. The Troemner™ 2-Prong Dual Adjust Nickel-Plated Zinc Clamp allows placement of apparatus at various distances from lab-frames without compromising the integrity of experiments.
  • Large grip adjustment range
  • 2 prong construction
  • Single or dual adjust
  • Nickel-plated zinc
TRUSTED_SUSTAINABILITY
Specifications
Type 2-Prong Dual Adjust Clamp
Material Nickel-plated Zinc