Learn More
Troemner™ Talboys™ All-Position Clamp Holder
Catalog No. 02300222 Shop All Troemner Products
Change view
Click to view available options
Type:
Channel Connector
Catalog No. Type
02-300-222 Channel Connector
1 options
Catalog No. 02-300-222 Supplier Troemner™ Supplier No. 30392248
Add to Cart
Add to Cart
Surpasses standard holding capabilities. The The Troemner™ Talboys™ All-Position Clamp Holder permits adjustments at any angle, in any plane.
  • Nickel-plated zinc construction
  • Holders are set at 90° to each other and connected by a 90°Connector, allowing 360° rotation
TRUSTED_SUSTAINABILITY
Specifications
Type Channel Connector