Learn More
Troemner™ Talboys™ Dimpled Mat
Catalog No. 02218017 Shop All OHAUS Products
Catalog No. 02-218-017 Supplier Troemner™ Supplier No. 30531235
Designed to hold centrifuge tubes, vials, culture tubes, and micro tubes securely in place. The Troemner™ Talboys™ Talboys™ Dimpled Mat is used with rocking and waving shakers.
    • Can be easily removed for cleaning and transporting of tubes from bench to tray
  • TRUSTED_SUSTAINABILITY