Learn More
Troemner™ Talboys™ Dimpled Mat
Catalog No. 02218019 Shop All OHAUS Products
Change view
Click to view available options
For Use With (Equipment):
1000RS Rocking Shaker
Catalog No. For Use With (Equipment)
02-218-019 1000RS Rocking Shaker
1 options
Catalog No. 02-218-019 Supplier Troemner™ Supplier No. 30531237
Designed to hold centrifuge tubes, vials, culture tubes, and micro tubes securely in place. The Troemner™ Talboys™ Talboys™ Dimpled Mat is used with rocking and waving shakers.
    • Can be easily removed for cleaning and transporting of tubes from bench to tray
  • TRUSTED_SUSTAINABILITY
    Specifications
    For Use With (Equipment) 1000RS Rocking Shaker