Learn More
Troemner™ Talboys™ Dual Adjust 2-Prong Nickel-Plated Zinc Clamp
Catalog No. 02216210 Shop All Troemner Products
Change view
Click to view available options
Material:
Nickel-plated Zinc
Catalog No. Material
02-216-210 Nickel-plated Zinc
1 options
Catalog No. 02-216-210 Supplier Troemner™ Supplier No. 30392209
Add to Cart
Add to Cart
Holds laboratory glassware and apparatus securely. The Troemner™ Talboys™ Dual Adjust 2-Prong Nickel-Plated Zinc Clamp offers easy positioning in the deepest fume hoods.
  • Large grip adjustment range
  • 2 prong construction
  • Single or dual adjust
  • Nickel-plated zinc
  • Constructed with round extension arms
  • Supplied with non-slip vinyl sleeves
  • TRUSTED_SUSTAINABILITY
    Specifications
    Type 2-Prong Dual Adjust Clamp
    Material Nickel-plated Zinc