Learn More
Troemner™ Talboys™ Fixed Position 2-Prong Single Adjust Nickel-Plated Zinc Clamp
Catalog No. 02300212 Shop All Troemner Products
Change view
Click to view available options
Material:
Nickel-plated Zinc
Catalog No. Material
02-300-212 Nickel-plated Zinc
1 options
Catalog No. 02-300-212 Supplier Troemner™ Supplier No. 30392218
Add to Cart
Add to Cart
Used to hold apparatus near the lab-frame where no adjustment is required after set-up. The Troemner™ Talboys™ Fixed Position 2-Prong Single Adjust Nickel-Plated Zinc Clamp is made in the USA.
  • Built-in holder grips rods up to 19mm (0.75in.) in diameter
  • Integral holder that can be rotated after attachment
  • Two prongs
  • Nickel-plated zinc construction
  • With non-slip vinyl sleeves
  • For temperatures above 100°C, fiberglass covers are included
TRUSTED_SUSTAINABILITY
Specifications
Type Fixed Position 2-Prong Single Adjust Clamp
Material Nickel-plated Zinc
Includes With non-slip vinyl sleeve.