Learn More
Troemner™ Talboys™ Heavy Duty Holder
Catalog No. 02216354 Shop All Troemner Products
Change view
Click to view available options
Type:
Heavy Duty Holder
Catalog No. Type
02-216-354 Heavy Duty Holder
1 options
Catalog No. 02-216-354 Supplier Troemner™ Supplier No. 30400045
Add to Cart
Add to Cart
Comprised of a sturdy aluminum alloy with a brushed finish. The Troemner™ Talboys™ Heavy Duty Holder is made in the USA.
  • Accommodates all shafts 1/4in. (6.4mm) to 1in. (25.4mm) diameter
  • Oversized, easy to grip knobs for precise tightening
  • Supports rods on a 4in. (10.2cm) long surface to prevent vibrations
TRUSTED_SUSTAINABILITY
Specifications
Type Heavy Duty Holder
Description Accommodates all shafts 1/4 in. (6.4 mm) to 1 in. (25.4 mm) diameter