Learn More
Troemner™ Talboys™ PVC Erlenmeyer Flask Clamp
Catalog No. 02218026 Shop All OHAUS Products
Change view
Click to view available options
Holds:
125 mL Erlenmeyer Flask
Catalog No. Holds
02-218-026 125 mL Erlenmeyer Flask
1 options
Catalog No. 02-218-026 Supplier Troemner™ Supplier No. 30531468
Manufactured to the highest standards. The Troemner™ Talboys™ PVC Erlenmeyer Flask Clamp is autoclavable.
  • Autoclavable
  • One piece, molded PVC construction
  • Will not scratch or mark flask like other clamps
  • Includes hardware for easy attachment to platforms
  • Attaching and removing of flasks is quick and easy
TRUSTED_SUSTAINABILITY
Specifications
Holds 125 mL Erlenmeyer Flask