Learn More
Troemner™ Talboys™ S-Connector, Nickel-Plated Zinc
Catalog No. 02216259 Shop All Troemner Products
Change view
Click to view available options
Type:
S-Connector
Catalog No. Type
02-216-259 S-Connector
1 options
Catalog No. 02-216-259 Supplier Troemner™ Supplier No. 30392257
Add to Cart
Add to Cart
Manufactured to the highest standards. The Troemner™ Talboys™ S-Connector, Nickel-Plated Zinc is ideal for supports requiring the connection of two perpendicular rods.
  • Connects two 13mm (0.51 in.) rods at 90° angle
  • Features separate adjustment screws for each rod location
  • Nickel-plated zinc
TRUSTED_SUSTAINABILITY
Specifications
Type S-Connector
Description Connects two 13 mm (0.51 in.) rods (at 90° angle)