Learn More
Troemner™ Talboys™ Stacking Tray
Catalog No. 02218014 Shop All OHAUS Products
Catalog No. 02-218-014 Supplier Troemner™ Supplier No. 30531229
Manufactured to the highest standards. The Troemner™ Talboys™ Stacking Tray attaches to shaker unit's included tray to add a second tier for higher capacity applications.
  • Easily attaches to the unit's included tray
  • Second tier tray mounts 3.5 in. (8.9cm) above lower tray
  • Includes hardware and a rubber mat
TRUSTED_SUSTAINABILITY
Specifications
Product Type Stacking Tray
Dimensions (L x W x H) 12.75 x 10 x 3.5 in. (32.4 x 25.4 x 8.9 cm)
Includes Hardware and Rubber Mat