Learn More
Troemner™ Talboys™ Stacking Tray
Catalog No. 19216378 Shop All Troemner Products
Catalog No. 19-216-378 Supplier Troemner™ Supplier No. 30390783
Add to Cart
Add to Cart
Manufactured to the highest standards. The Troemner™ Talboys™ Stacking Tray attaches to shaker unit's included tray to add a second tier for higher capacity applications.
  • Includes hardware and a rubber mat
TRUSTED_SUSTAINABILITY
Specifications
Product Type Stacking Tray
Length (English) 12.75 in.
Length (Metric) 32.4 cm
Width (English) 10 in.
For Use With (Equipment) Advanced 1000RS Rocking Shaker