Learn More
Troemner™ Talboys™ Stainless Steel Erlenmeyer Flask Clamp
Catalog No. 02218022 Shop All OHAUS Products
Change view
Click to view available options
Holds:
25 mL Erlenmeyer Flask
Catalog No. Holds
02-218-022 25 mL Erlenmeyer Flask
1 options
Catalog No. 02-218-022 Supplier Troemner™ Supplier No. 30531220
Manufactured to the highest standards. The Troemner™ Talboys™ Stainless Steel Erlenmeyer Flask Clamp keeps flasks secure.
  • Designed to hold Erlenmeyer flasks
  • Constructed of type 302 and 304 stainless steel
  • Includes hardware for easy attachment to platforms
  • Flask clamps are supplied with a spring to hold flask in place
TRUSTED_SUSTAINABILITY
Specifications
Holds 25 mL Erlenmeyer Flask
Material Stainless Steel