Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS™ Support Rod and Clamp Kit
SCI_ED

Catalog No. S38771 Shop All OHAUS Products
Change view
Click to view available options
Color:
Silver
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S38771 Silver
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S38771 Supplier OHAUS™ Supplier No. 30400146/EMD
Add to Cart
Add to Cart

For holding temperature probes, thermometers or other glass tubes.

  • For all Hotplates or Hotplate-Stirrers
  • Includes an 45.7cm (18 in.), Stainless-steel support rod, thermometer/temperature probe extension clamp, 3-prong medium swivel clamp and hook connector

Compliance

UL, cUL, CE marked

TRUSTED_SUSTAINABILITY

Specifications

Quantity Ea.
Type Clamp Kit
Material Nickel-plated
Includes 18 in. support rod, thermometer/temperature probe extension clamp, 3 prong medium swivel clamp and hook connector
For Use With (Equipment) Hotplates, stirrers and hotplate and stirrers
Description Support Rod And Clamp Kit
Color Silver
Jaw Type 3-Prong Medium Swivel Clamp
Diameter (Metric) Inside 0 to 69 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.