Learn More
Troemner™ Talboys™ Swivel Holder, Nickel-Plated Zinc
Catalog No. 02214150 Shop All Troemner Products
Change view
Click to view available options
Type:
Swivel Holder
Catalog No. Type
02-214-150 Swivel Holder
1 options
Catalog No. 02-214-150 Supplier Troemner™ Supplier No. 30392213
Add to Cart
Add to Cart
Manufactured to the highest standards. The Troemner™ Talboys™ Swivel Holder, Nickel-Plated Zinc allows close proximity between items being held.
  • Two rod holders with center swivel capacity allow tilting of clamps at any angle in parallel planes
  • Outside adjustments screw allows close proximity between items being held
  • Nickel-plated zinc construction
TRUSTED_SUSTAINABILITY
Specifications
Type Swivel Holder
Description Nickel-plated zinc construction