Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Talfirastide acetate | 2855063-75-9 | 99.8% | 959.06 | C43H66N12O13 | 10MM 1ML
SDP

Catalog No. 5000238933
Encompass_Preferred

Talfirastide acetate is the acetate salt of the endogenous heptapeptide angiotensin-(1-7), used in research on the renin-angiotensin system and studies of cardiovascular, anti-inflammatory, and anti-fibrotic pathways. It is provided as a 10 mM solution (1 mL) or as a powder for reconstitution; sequence is Asp-Arg-Val-Tyr-Ile-His-Pro (DRVYIHP).

  • Heptapeptide angiotensin-(1-7) acetate (sequence DRVYIHP).
  • Supplied as 10 mM, 1 mL solution; powder available for reconstitution.
  • Molecular weight 959.06; chemical formula C43H66N12O13.
  • Purity reported as 99.8%.
  • Storage: powder sealed at -80°C (2 years) or -20°C (1 year); in solvent -80°C (6 months) or -20°C (1 month).
  • Intended for research use only.

Catalog No. 50-002-38933 Supplier Medchemexpress LLC Supplier No. HY12403A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Talfirastide acetate is the acetate salt of the endogenous heptapeptide angiotensin-(1-7), used in research on the renin-angiotensin system and studies of cardiovascular, anti-inflammatory, and anti-fibrotic pathways. It is provided as a 10 mM solution (1 mL) or as a powder for reconstitution; sequence is Asp-Arg-Val-Tyr-Ile-His-Pro (DRVYIHP).

  • Heptapeptide angiotensin-(1-7) acetate (sequence DRVYIHP).
  • Supplied as 10 mM, 1 mL solution; powder available for reconstitution.
  • Molecular weight 959.06; chemical formula C43H66N12O13.
  • Purity reported as 99.8%.
  • Storage: powder sealed at -80°C (2 years) or -20°C (1 year); in solvent -80°C (6 months) or -20°C (1 month).
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.