Learn More
Medchemexpress LLC Talibegron hydrochloride (ZD2079 hydrochloride) | 178600-17-4 | C18H22ClNO4 | 1 MG
SDP
Catalog No. 5000376407
Encompass_Preferred
Talibegron hydrochloride (ZD2079 hydrochloride) is a potent β3-adrenoceptor agonist with a pD2 of 3.72 on phenylephrine-preconstricted rat mesenteric artery. It has a potent vasorelaxant effect.
  • Potent β3-adrenoceptor agonist.
  • Has potent vasorelaxant effect.
  • Produces a concentration-dependent relaxation of phenylephrine-preconstricted isolated mesenteric arteries.
  • Causes full relaxation with 1 mM.
Catalog No. 50-003-76407 Supplier Medchemexpress LLC Supplier No. HY153781MG
Talibegron hydrochloride (ZD2079 hydrochloride) is a potent β3-adrenoceptor agonist with a pD2 of 3.72 on phenylephrine-preconstricted rat mesenteric artery. It has a potent vasorelaxant effect.
  • Potent β3-adrenoceptor agonist.
  • Has potent vasorelaxant effect.
  • Produces a concentration-dependent relaxation of phenylephrine-preconstricted isolated mesenteric arteries.
  • Causes full relaxation with 1 mM.