Learn More
Cayman Chemical -TalInolol 10mg
SDP
Catalog No. 502971859
Encompass_Preferred
A β1-selective adrenoceptor antagonist well known for its cardioprotective and antihypertensive activity; often used to examine the activity of the drug-transporting MDR1 gene product P-glycoprotein
Catalog No. 50-297-1859 Supplier Cayman Chemical Supplier No. 1611610 MG
A β1-selective adrenoceptor antagonist well known for its cardioprotective and antihypertensive activity; often used to examine the activity of the drug-transporting MDR1 gene product P-glycoprotein