Learn More
Medchemexpress LLC HY-A0008 10mg Medchemexpress, Talipexole dihydrochloride CAS:36085-73-1 Purity:>98%
SDP
Catalog No. 502030995
Encompass_Preferred
Medchemexpress, HY-A0008 10mg Talipexole dihydrochloride CAS:36085-73-1 Talipexole dihydrochloride (B-HT 920 dihydrochloride) is a dopamine D2 receptor agonist, α2-adrenoceptor agonist and 5-HT3 receptor antagonist, which displays antiParkinsonian activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0995 Supplier Medchemexpress LLC Supplier No. HYA000810MG
Medchemexpress, HY-A0008 10mg Talipexole dihydrochloride CAS:36085-73-1 Talipexole dihydrochloride (B-HT 920 dihydrochloride) is a dopamine D2 receptor agonist, α2-adrenoceptor agonist and 5-HT3 receptor antagonist, which displays antiParkinsonian activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.