Learn More
Medchemexpress LLC Talipexole dihydrochloride | 36085-73-1 | 99.8% | C10H17Cl2N3S | 1 ML
SDP
Catalog No. 5000433329
Encompass_Preferred
Talipexole dihydrochloride is a dopamine D2 receptor agonist, α2-adrenoceptor agonist, and 5-HT3 receptor antagonist, which exhibits anti-Parkinsonian activity.
  • Ready for reconstitution
  • Displays anti-Parkinsonian activity
Catalog No. 50-004-33329 Supplier Medchemexpress LLC Supplier No. HYA000810MM/1ML
Talipexole dihydrochloride is a dopamine D2 receptor agonist, α2-adrenoceptor agonist, and 5-HT3 receptor antagonist, which exhibits anti-Parkinsonian activity.
  • Ready for reconstitution
  • Displays anti-Parkinsonian activity