Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Talipexole (dihydrochloride) | 36085-73-1 | 99.8% | C10H17Cl2N3S | 25 MG
SDP

Catalog No. 5000357535
Encompass_Preferred

Talipexole dihydrochloride is a dopamine D2 receptor agonist, α2-adrenoceptor agonist, and 5-HT3 receptor antagonist. It displays anti-Parkinsonian activity and is a research chemical used in studies related to neurological and neurodegenerative diseases.

  • Dopamine D2 receptor agonist.
  • α2-adrenoceptor agonist.
  • 5-HT3 receptor antagonist.
  • Exhibits anti-Parkinsonian activity.
  • Targets D2 Receptor (IC50: 25 nM), 5-HT3 Receptor, and α2-adrenergic receptor.
  • For research use only.

Catalog No. 50-003-57535 Supplier Medchemexpress LLC Supplier No. 25MGHYA0008
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Talipexole dihydrochloride is a dopamine D2 receptor agonist, α2-adrenoceptor agonist, and 5-HT3 receptor antagonist. It displays anti-Parkinsonian activity and is a research chemical used in studies related to neurological and neurodegenerative diseases.

  • Dopamine D2 receptor agonist.
  • α2-adrenoceptor agonist.
  • 5-HT3 receptor antagonist.
  • Exhibits anti-Parkinsonian activity.
  • Targets D2 Receptor (IC50: 25 nM), 5-HT3 Receptor, and α2-adrenergic receptor.
  • For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.