Learn More
California Pacific Lab & Consulting TALKING ELECTRONIC LIQUID
SDP
Catalog No. NC2080539
Encompass_Preferred
talking electronic liquid level indicator for overflow prevention.Features and BenefitsEasy to set up and maintainPrevents overflowsUL tested for non-sparkingGreat for use on HPLC waste containersWorks on glass and plastic containersFits containers as small as 2L and as large as 40 gallon*includes 8' USB cable (300V) - USB power means there is no battery to fail and cause a spillSpecifications2" diameter x 2.5" across1.7 cm thickABS plastic outerFits maximum container circumference: 54.5" (17.35" diameter / approximately 30-40 gallon)*Fits minimum container circumference: 14.75" (4.7" diameter / approximately 2 Liter)Elastic band stretches up to 27" (relaxed) and 54.5" (stretched)
Catalog No. NC2080539 Supplier California Pacific Lab & Consulting Supplier No. CPTELLI01
talking electronic liquid level indicator for overflow prevention.Features and BenefitsEasy to set up and maintainPrevents overflowsUL tested for non-sparkingGreat for use on HPLC waste containersWorks on glass and plastic containersFits containers as small as 2L and as large as 40 gallon*includes 8' USB cable (300V) - USB power means there is no battery to fail and cause a spillSpecifications2" diameter x 2.5" across1.7 cm thickABS plastic outerFits maximum container circumference: 54.5" (17.35" diameter / approximately 30-40 gallon)*Fits minimum container circumference: 14.75" (4.7" diameter / approximately 2 Liter)Elastic band stretches up to 27" (relaxed) and 54.5" (stretched)