Learn More
Electron Microscopy Sciences Talking Timer
SDP
Catalog No. 50949422
Encompass_Preferred
A unique timer featuring a human voice that makes announcements which can be heard from any where in the lab. In the count-down mode, it will announce the remaining time every hour. In the final hour, it will announce the time remaining every 10 mins, every min in the final 10 mins, and every sec in the final 10 secs. It has a choice of 6 distinctive final alarm sounds. In the dark, in the clock mode, you can touch any button and the time will be announced. The voice may be turned off if desired. Other features include a large 3/4" display with icons to indicate various modes. You are able to recall the previous count-down setting for immediate reuse and memorizes other frequently used setting for reuse at any time. Counts up or down to 24 hours and has a built in stand for bench top use. It clips to a pocket or sticks to any metal surface with a magnetic back. Button cell batteries are included. The timer also available with a N.I.S.T. traceable certificate of compliance.
Catalog No. 50-949-422 Supplier Electron Microscopy Sciences Supplier No. 7260101
A unique timer featuring a human voice that makes announcements which can be heard from any where in the lab. In the count-down mode, it will announce the remaining time every hour. In the final hour, it will announce the time remaining every 10 mins, every min in the final 10 mins, and every sec in the final 10 secs. It has a choice of 6 distinctive final alarm sounds. In the dark, in the clock mode, you can touch any button and the time will be announced. The voice may be turned off if desired. Other features include a large 3/4" display with icons to indicate various modes. You are able to recall the previous count-down setting for immediate reuse and memorizes other frequently used setting for reuse at any time. Counts up or down to 24 hours and has a built in stand for bench top use. It clips to a pocket or sticks to any metal surface with a magnetic back. Button cell batteries are included. The timer also available with a N.I.S.T. traceable certificate of compliance.