Learn More
Research Products International Corp Talking Timer
SDP
Catalog No. 501978427
Encompass
A unique and very useful timer featuring a clear pleasant human voice. Eliminate the need to continually visually check time remaining via voice announcements that can be heard from anywhere in the lab.
In the count-down mode Talking Timer will announce the remaining time every hour. In the final hour it will announce the time remaining every 10 minutes, every minute in the final 10 minutes, and every second in the final 10 seconds.
Other features include a large 3/4" display with icons to indicate various modes, such as voice off/on, count-down/count-up, etc. Recalls the previous count-down setting for immediate reuse and memorizes another frequently used setting for reuse at any time. Counts up or down to 24 hours. Both functions and the clock work simultaneously.
Button cell (AG13) batteries are included.
5 Year Warranty
Catalog No. 50-197-8427 Supplier Research Products International Corp Supplier No. 810038
A unique and very useful timer featuring a clear pleasant human voice. Eliminate the need to continually visually check time remaining via voice announcements that can be heard from anywhere in the lab.
In the count-down mode Talking Timer will announce the remaining time every hour. In the final hour it will announce the time remaining every 10 minutes, every minute in the final 10 minutes, and every second in the final 10 seconds.
Other features include a large 3/4" display with icons to indicate various modes, such as voice off/on, count-down/count-up, etc. Recalls the previous count-down setting for immediate reuse and memorizes another frequently used setting for reuse at any time. Counts up or down to 24 hours. Both functions and the clock work simultaneously.
Button cell (AG13) batteries are included.
5 Year Warranty