Learn More
MSC Corrugated Shipping Box 6x6x40 in., 1 ea
Catalog No. 501915449
Encompass
Tall corrugated boxes are designed for shipping irregularly shaped items. Featuring an end-loading design, these boxes offer easy insertion and sealing, and can serve as an alternative to mailing tubes.
  • Manufactured from professional grade 200#/ECT-32 C kraft corrugated
  • Durable construction supports up to 65 lb.
  • Sold in bundle quantities for convenience
  • Ships flat to save on storage space
  • Reusable and recyclable
  • Made in the USA
Catalog No. 50-191-5449 Supplier MSC Supplier No. 39549472
Tall corrugated boxes are designed for shipping irregularly shaped items. Featuring an end-loading design, these boxes offer easy insertion and sealing, and can serve as an alternative to mailing tubes.
  • Manufactured from professional grade 200#/ECT-32 C kraft corrugated
  • Durable construction supports up to 65 lb.
  • Sold in bundle quantities for convenience
  • Ships flat to save on storage space
  • Reusable and recyclable
  • Made in the USA