Learn More
Mitegen Tall Foam Dewar, 1800 mL Capacity, Lightweight, Shatterproof, Excellent Thermal Insulation for Lab Chilling
SDP
Catalog No. 501187324
Encompass
This Tall Foam Dewar offers a versatile and safe solution for various laboratory applications demanding precise temperature maintenance for sensitive materials. With a generous 1800 mL (1.8 Liter) capacity, this dewar is ideally suited for handling liquid nitrogen, dry ice, ice, or other cooling agents, particularly when deeper immersion of samples or tools is required. Constructed from robust, high-density foam, it provides excellent thermal insulation, crucial for maintaining stable temperatures and ensuring the integrity of your samples or reagents. Its lightweight and inherently shatterproof design enhances safety in the lab, minimizing risks associated with accidental drops compared to traditional glass vessels. Featuring a wide, accessible opening and a stable base, this foam dewar is a reliable and indispensable addition for quick chilling, temporary cryogenic storage, and other temperature-sensitive processes.
Catalog No. 50-118-7324 Supplier Mitegen Supplier No. MTD1800
This Tall Foam Dewar offers a versatile and safe solution for various laboratory applications demanding precise temperature maintenance for sensitive materials. With a generous 1800 mL (1.8 Liter) capacity, this dewar is ideally suited for handling liquid nitrogen, dry ice, ice, or other cooling agents, particularly when deeper immersion of samples or tools is required. Constructed from robust, high-density foam, it provides excellent thermal insulation, crucial for maintaining stable temperatures and ensuring the integrity of your samples or reagents. Its lightweight and inherently shatterproof design enhances safety in the lab, minimizing risks associated with accidental drops compared to traditional glass vessels. Featuring a wide, accessible opening and a stable base, this foam dewar is a reliable and indispensable addition for quick chilling, temporary cryogenic storage, and other temperature-sensitive processes.