Learn More
Cole-Parmer Cole-Parmer Tall-Form Crucible, High Alumina, 250 mL, 1/Ea
Supplier:  Cole-Parmer 6310108
Encompass_Preferred
Unglazed tall-form crucibles are 99.7% pure aluminum oxide. Withstand temperatures up to 3182°F (1750°C). High alumina labware temperature change rate should not exceed 150°C/hour. Autoclavable
Catalog No. NC1700303