Learn More
ATCC TALL-104; T Cell Leukemia(ALL); Human
Supplier:  ATCC CRL11386
Encompass
TALL-104 is a T lymphoblast cell line that was established from the peripheral blood of a two-year-old male child in relapse with T-ALL. Applications include immune system disorder, immunology, and immuno-oncology research.
Catalog No. 50-238-3482