Learn More
TARGETMOL CHEMICALS INC TALNIFLUMATE 25MG
SDP
Catalog No. 502579874
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Talniflumate (Somalgen) is a calcium-activated chloride channel (hCLCA1/mCLCA3) blocker that reduces mucin synthesis and releases in animal models and cell culture. It possesses anti-inflammatory actions via inhibition of Cox (cyclooxygenase) and inhibits Cl-/HCO3- exchange activity. It also increases survival in a cystic fibrosis mouse model of the distal intestinal obstructive syndrome. purity: 99%
Catalog No. 50-257-9874 Supplier TARGETMOL CHEMICALS INC Supplier No. T450325MG
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Talniflumate (Somalgen) is a calcium-activated chloride channel (hCLCA1/mCLCA3) blocker that reduces mucin synthesis and releases in animal models and cell culture. It possesses anti-inflammatory actions via inhibition of Cox (cyclooxygenase) and inhibits Cl-/HCO3- exchange activity. It also increases survival in a cystic fibrosis mouse model of the distal intestinal obstructive syndrome. purity: 99%