Learn More
Cayman Chemical Talniflumate, Purity: ≥95%, CAS: 66898-62-2, 50 mg
SDP
Catalog No. 502991635
Encompass_Preferred
A prodrug form of niflumic acid; decreases the rate of chloride/bicarbonate exchange in isolated mouse intestinal villous epithelial cells at 100 µM; dietary administration increases survival but has no effect on the number of crypt goblet cells in the jejunum or cecum in a cystic fibrosis mouse model of distal intestinal obstructive syndrome at 88 mg/kg
Catalog No. 50-299-1635 Supplier Cayman Chemical Supplier No. 2962350 MG
A prodrug form of niflumic acid; decreases the rate of chloride/bicarbonate exchange in isolated mouse intestinal villous epithelial cells at 100 µM; dietary administration increases survival but has no effect on the number of crypt goblet cells in the jejunum or cecum in a cystic fibrosis mouse model of distal intestinal obstructive syndrome at 88 mg/kg