Learn More
Medchemexpress LLC Talniflumate | 66898-62-2 | MFCD00866135 | 99.7% | 414.33 g/mol | C21H13F3N2O4 | 10 MG
SDP
Catalog No. 5000244853
Encompass_Preferred
Talniflumate is a small-molecule Ca2+-activated chloride channel (CaCC) blocker with reported analgesic and anti-inflammatory activity, used as a research reagent in studies of mucin regulation and inflammation. It is supplied as solids and as a DMSO solution for assay flexibility.
  • CaCC blocker with reported analgesic and anti-inflammatory activity.
  • Supplied as solid packages and as a DMSO solution for assay flexibility.
  • High purity suitable for research applications.
  • Molecular weight 414.33 g/mol and formula C21H13F3N2O4.
  • Applicable to mucin regulation and cystic fibrosis research models.
Catalog No. 50-002-44853 Supplier Medchemexpress LLC Supplier No. HY10337010MG
Talniflumate is a small-molecule Ca2+-activated chloride channel (CaCC) blocker with reported analgesic and anti-inflammatory activity, used as a research reagent in studies of mucin regulation and inflammation. It is supplied as solids and as a DMSO solution for assay flexibility.
  • CaCC blocker with reported analgesic and anti-inflammatory activity.
  • Supplied as solid packages and as a DMSO solution for assay flexibility.
  • High purity suitable for research applications.
  • Molecular weight 414.33 g/mol and formula C21H13F3N2O4.
  • Applicable to mucin regulation and cystic fibrosis research models.