Learn More
Medchemexpress LLC Talniflumate | 66898-62-2 | MFCD00866135 | 99.7% | 414.33 g/mol | C21H13F3N2O4 | 5 MG
SDP
Catalog No. 5000245207
Encompass_Preferred
Talniflumate is a research chemical prodrug of niflumic acid that is converted in vivo by esterases and acts as an orally active Ca2+-activated chloride channel (CaCC) blocker with reported analgesic and anti-inflammatory effects in preclinical models.
  • Prodrug of niflumic acid converted by esterases in vivo.
  • Orally active Ca2+-activated chloride channel (CaCC) blocker.
  • Suitable for in vitro and in vivo preclinical studies.
  • Powder form with recommended storage at -20°C for long-term stability.
  • Soluble in DMSO (≈62.5 mg/mL) and formulatable for in vivo dosing.
Catalog No. 50-002-45207 Supplier Medchemexpress LLC Supplier No. HY1033705MG
Talniflumate is a research chemical prodrug of niflumic acid that is converted in vivo by esterases and acts as an orally active Ca2+-activated chloride channel (CaCC) blocker with reported analgesic and anti-inflammatory effects in preclinical models.
  • Prodrug of niflumic acid converted by esterases in vivo.
  • Orally active Ca2+-activated chloride channel (CaCC) blocker.
  • Suitable for in vitro and in vivo preclinical studies.
  • Powder form with recommended storage at -20°C for long-term stability.
  • Soluble in DMSO (≈62.5 mg/mL) and formulatable for in vivo dosing.