Learn More
Medchemexpress LLC Talniflumate | 66898-62-2 | MFCD00866135 | 99.7% | 414.33 g/mol | C21H13F3N2O4 | 50 MG
SDP
Catalog No. 5000244614
Encompass_Preferred
Talniflumate is a research-grade calcium-activated chloride channel (CaCC) blocker and a proagent of niflumic acid used in pharmacological and physiological studies. It is supplied as a high-purity solid suitable for preparation of stock solutions for in vitro and in vivo experiments.
  • CaCC blocker and proagent of niflumic acid
  • High reported purity for reliable experimental results
  • Soluble in DMSO at high concentrations for stock preparation
  • Compatible with in vivo formulations at specified concentrations
  • Stable when stored frozen as a powder for extended periods
Catalog No. 50-002-44614 Supplier Medchemexpress LLC Supplier No. HY10337050MG
Talniflumate is a research-grade calcium-activated chloride channel (CaCC) blocker and a proagent of niflumic acid used in pharmacological and physiological studies. It is supplied as a high-purity solid suitable for preparation of stock solutions for in vitro and in vivo experiments.
  • CaCC blocker and proagent of niflumic acid
  • High reported purity for reliable experimental results
  • Soluble in DMSO at high concentrations for stock preparation
  • Compatible with in vivo formulations at specified concentrations
  • Stable when stored frozen as a powder for extended periods