Learn More
Takara Bio TALON® Metal Affinity Resin 10 ml
SDP
Catalog No. NC9436748
Encompass
Immobilized metal affinity chromatography resin for the purification of recombinant his-tagged proteins under native or denaturing conditions. 10 ml
Catalog No. NC9436748 Supplier Takara Bio Supplier No. 635501
Immobilized metal affinity chromatography resin for the purification of recombinant his-tagged proteins under native or denaturing conditions. 10 ml