Learn More
Takara Bio TALON® Metal Affinity Resin 250 ml
SDP
Catalog No. NC9499432
Encompass
Immobilized metal affinity chromatography resin for the purification of recombinant his-tagged proteins under native or denaturing conditions. 250 ml
Catalog No. NC9499432 Supplier Takara Bio Supplier No. 635504
Immobilized metal affinity chromatography resin for the purification of recombinant his-tagged proteins under native or denaturing conditions. 250 ml