Learn More
Takara Bio TALON® Metal Affinity Resin 25 ml
SDP
Catalog No. NC9306569
Encompass
Immobilized metal affinity chromatography resin for the purification of recombinant his-tagged proteins under native or denaturing conditions. 25 ml
Catalog No. NC9306569 Supplier Takara Bio Supplier No. 635502
Immobilized metal affinity chromatography resin for the purification of recombinant his-tagged proteins under native or denaturing conditions. 25 ml