Learn More
Takara Bio TALON SPRFLW AFNTY RESIN 100ML
SDP
Catalog No. NC9552006
Encompass
TALON Superflow Metal Affinity Resin, 100mL, For large scale, medium pressure purification of 6xHis-tagged biomolecules, Exceptional performance under native of denaturing conditions, Minimal metal leaching
Catalog No. NC9552006 Supplier Takara Bio Supplier No. 635507
TALON Superflow Metal Affinity Resin, 100mL, For large scale, medium pressure purification of 6xHis-tagged biomolecules, Exceptional performance under native of denaturing conditions, Minimal metal leaching