Learn More
TARGETMOL CHEMICALS INC TALOTREXIN AMMONIUM 5MG
SDP
Catalog No. 502579381
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Talotrexin ammonium is a non-polyglutamic acid antifolate. Talotrexin ammonium inhibits tumor growth by targeting dihydrofolate reductase (DHFR) thereby improving anti-tumor activity in a wide range of cancer models helping to inhibit tumor growth. purity: 97%
Catalog No. 50-257-9381 Supplier TARGETMOL CHEMICALS INC Supplier No. T642955MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Talotrexin ammonium is a non-polyglutamic acid antifolate. Talotrexin ammonium inhibits tumor growth by targeting dihydrofolate reductase (DHFR) thereby improving anti-tumor activity in a wide range of cancer models helping to inhibit tumor growth. purity: 97%