Learn More
Medchemexpress LLC Talquetamab (Synonyms: JNJ-64407564) 1mg | CAS No. : 2226212-40-2
SDP
Catalog No. NC3355875
Encompass_Preferred
Talquetamab (JNJ-64407564) is a humanized bispecific antibody that binds to GPRC5D (member of G protein-coupled receptor family C5 group D) and CD3 to induce T cell-mediated killing of GPRC5D-expressing MM cells through T cell recruitment and activation. Talquetamab (JNJ-64407564) has antitumor activity.
Catalog No. NC3355875 Supplier Medchemexpress LLC Supplier No. HYP993941
Talquetamab (JNJ-64407564) is a humanized bispecific antibody that binds to GPRC5D (member of G protein-coupled receptor family C5 group D) and CD3 to induce T cell-mediated killing of GPRC5D-expressing MM cells through T cell recruitment and activation. Talquetamab (JNJ-64407564) has antitumor activity.