Learn More
Medchemexpress LLC Taltirelin acetate | 1549593-23-8 | 99.4% | 1 ML
SDP
Catalog No. 5000408779
Encompass_Preferred
Taltirelin acetate (TA-0910) is an acetate form of Taltirelin, an orally effective analogue of thyrotropin releasing hormone (TRH) and a TRH receptor superagonist. It can cross the blood-brain barrier and stimulates an increase in cytosolic Ca2+ concentration. This compound has neuroprotective effects in both cellular and animal models of Parkinson's disease and helps alleviate fatigue-like behavior in mouse models of cancer-related fatigue.
  • Increases cell viability
  • Reduces apoptosis in SH-SY5Y cells
  • Inhibits tau phosphorylation and AEP activation
  • Improves locomotor function
  • Preserves dopaminergic neurons in the substantia nigra
Catalog No. 50-004-08779 Supplier Medchemexpress LLC Supplier No. HYB0596A10MM/1ML
Taltirelin acetate (TA-0910) is an acetate form of Taltirelin, an orally effective analogue of thyrotropin releasing hormone (TRH) and a TRH receptor superagonist. It can cross the blood-brain barrier and stimulates an increase in cytosolic Ca2+ concentration. This compound has neuroprotective effects in both cellular and animal models of Parkinson's disease and helps alleviate fatigue-like behavior in mouse models of cancer-related fatigue.
  • Increases cell viability
  • Reduces apoptosis in SH-SY5Y cells
  • Inhibits tau phosphorylation and AEP activation
  • Improves locomotor function
  • Preserves dopaminergic neurons in the substantia nigra