Learn More
Medchemexpress LLC HY-B0596A 5mg Medchemexpress, Taltirelin (acetate) CAS:1549593-23-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0596A5MG
Encompass_Preferred
Medchemexpress, HY-B0596A 5mg Taltirelin (acetate) CAS:1549593-23-8 Taltirelin acetate (TA-0910 acetate) is a superagonist at thyrotropin-releasing hormone receptor (TRH-R) with an IC 50 of 910 nM and EC 50 of 36 nM for stimulating an increase in cytosolic Ca 2+ concentration (Ca 2+ release) [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2924