Learn More
Medchemexpress LLC Taltobulin hydrochloride | 228266-40-8 | 99.3% | 510.11 Da | C27H44ClN3O4 | 100 MG
SDP
Catalog No. 5000204428
Encompass_Preferred
Taltobulin hydrochloride is a synthetic analogue of the tripeptide hemiasterlin and a potent antimicrotubule agent. It inhibits tubulin polymerization, disrupts microtubule organization, and induces mitotic arrest, and is supplied as a solid research reagent for in vitro and in vivo studies.
  • Potent tubulin inhibitor with low-nanomolar activity in tumor cell lines.
  • High purity suitable for research applications.
  • Supplied as a solid powder for solution preparation in DMSO or aqueous buffers.
  • Recommended solvent and formulation guidance provided for in vitro and in vivo use.
  • Store sealed at 4°C; solutions stable at -80°C for long-term storage.
Catalog No. 50-002-04428 Supplier Medchemexpress LLC Supplier No. HY15584B100MG
Taltobulin hydrochloride is a synthetic analogue of the tripeptide hemiasterlin and a potent antimicrotubule agent. It inhibits tubulin polymerization, disrupts microtubule organization, and induces mitotic arrest, and is supplied as a solid research reagent for in vitro and in vivo studies.
  • Potent tubulin inhibitor with low-nanomolar activity in tumor cell lines.
  • High purity suitable for research applications.
  • Supplied as a solid powder for solution preparation in DMSO or aqueous buffers.
  • Recommended solvent and formulation guidance provided for in vitro and in vivo use.
  • Store sealed at 4°C; solutions stable at -80°C for long-term storage.