Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Taltobulin (hydrochloride) | 228266-40-8 | 99.3% | 510.11 g/mol | C27H44ClN3O4 | 25 MG
SDP

Catalog No. 5000204448
Encompass_Preferred

Taltobulin (hydrochloride) is the hydrochloride salt of taltobulin (HTI-286), a synthetic analogue of the tripeptide hemiasterlin that acts as a potent tubulin inhibitor for research use.

  • hydrochloride salt form suitable for research applications.
  • potent tubulin inhibitor and antimicrotubule agent.
  • molecular weight 510.11 g/mol.
  • chemical formula C27H44ClN3O4.
  • high purity (99.3% by LC-MS).
  • physical form: white to off-white solid.

Catalog No. 50-002-04448 Supplier Medchemexpress LLC Supplier No. HY15584B25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Taltobulin (hydrochloride) is the hydrochloride salt of taltobulin (HTI-286), a synthetic analogue of the tripeptide hemiasterlin that acts as a potent tubulin inhibitor for research use.

  • hydrochloride salt form suitable for research applications.
  • potent tubulin inhibitor and antimicrotubule agent.
  • molecular weight 510.11 g/mol.
  • chemical formula C27H44ClN3O4.
  • high purity (99.3% by LC-MS).
  • physical form: white to off-white solid.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.