Learn More
Medchemexpress LLC Taltobulin intermediate-3 | 610786-69-1 | 99.7% | 235.75 | 100 MG
SDP
Catalog No. 5000404773
Encompass_Preferred
Taltobulin intermediate-3 is an intermediate in the synthesis of Taltobulin. Taltobulin is a common toxin component in ADC preparation (ADC Cytotoxin), and it is also a powerful tubulin (Microtubule/Tubulin) inhibitor.
  • Disrupts tubulin polymerization
  • Induces mitotic arrest
  • Induces apoptosis
Catalog No. 50-004-04773 Supplier Medchemexpress LLC Supplier No. HY41054100MG
Taltobulin intermediate-3 is an intermediate in the synthesis of Taltobulin. Taltobulin is a common toxin component in ADC preparation (ADC Cytotoxin), and it is also a powerful tubulin (Microtubule/Tubulin) inhibitor.
  • Disrupts tubulin polymerization
  • Induces mitotic arrest
  • Induces apoptosis