Learn More
Medchemexpress LLC Taltobulin trifluoroacetate | 228266-41-9 | 99.1% | 587.67 g/mol | C29H44F3N3O6 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY15584A10MM 1ML
Encompass_Preferred
Taltobulin trifluoroacetate is a tubulin polymerization inhibitor supplied as a 10 mM solution in DMSO for research use. It disrupts microtubule organization, induces mitotic arrest, and is used in cell-based and biochemical assays to study microtubule-targeting mechanisms. Not for human or clinical use.
  • Ready-to-use 10 mM solution in DMSO.
  • Reported purity of 99.1%.
  • Molecular formula C29H44F3N3O6; molecular weight 587.67 g/mol.
  • Inhibits tubulin polymerization and disrupts microtubule organization.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-04414