Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tam-in-2 (pyrrolotriazine compound 0904) | 2135642-56-5 | 99.9% | 583.59 g/mol | C31H27F2N7O3 | 100MG
SDP

Catalog No. 5000234981
Encompass_Preferred

TAM-IN-2 is a pyrrolotriazine-derived small-molecule inhibitor of TAM family receptor tyrosine kinases (described in patent US 20170275290 A1, compound 0904). Supplied as a high-purity solid for research use, it is intended for biochemical and cellular studies of TAM signaling and pathway modulation.

  • CAS number 2135642-56-5
  • Molecular weight 583.59 g/mol
  • Chemical formula C31H27F2N7O3
  • Purity 99.9%
  • Physical form solid; color white to light yellow
  • Solubility DMSO 5 mg/mL (may require sonication)
  • Storage: powder -20°C (stable 3 years); in solvent -80°C (2 years)
  • Available as a 100 mg laboratory package
  • Safety data sheet available in multiple languages

Catalog No. 50-002-34981 Supplier Medchemexpress LLC Supplier No. HY126216100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TAM-IN-2 is a pyrrolotriazine-derived small-molecule inhibitor of TAM family receptor tyrosine kinases (described in patent US 20170275290 A1, compound 0904). Supplied as a high-purity solid for research use, it is intended for biochemical and cellular studies of TAM signaling and pathway modulation.

  • CAS number 2135642-56-5
  • Molecular weight 583.59 g/mol
  • Chemical formula C31H27F2N7O3
  • Purity 99.9%
  • Physical form solid; color white to light yellow
  • Solubility DMSO 5 mg/mL (may require sonication)
  • Storage: powder -20°C (stable 3 years); in solvent -80°C (2 years)
  • Available as a 100 mg laboratory package
  • Safety data sheet available in multiple languages

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.