Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TAM557 TFA | 98.5% | 1119.34 | 1 MG
SDP

Catalog No. 5000391507
Encompass_Preferred

TAM557 TFA is a cytotoxic tubulysin compound designed for conjugation to transport vehicles such as proteins, peptides, small molecules, or polymeric carriers that incorporate a targeting principle. It is intended for research use only.

  • Cytotoxic tubulysin compound
  • Modified for conjugation to transport vehicles
  • Appearance: solid, white to off-white
  • Shipping at room temperature
  • Store at -20°C, sealed, away from moisture and light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-003-91507 Supplier Medchemexpress LLC Supplier No. HY160473A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TAM557 TFA is a cytotoxic tubulysin compound designed for conjugation to transport vehicles such as proteins, peptides, small molecules, or polymeric carriers that incorporate a targeting principle. It is intended for research use only.

  • Cytotoxic tubulysin compound
  • Modified for conjugation to transport vehicles
  • Appearance: solid, white to off-white
  • Shipping at room temperature
  • Store at -20°C, sealed, away from moisture and light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.